Learn More
Cayman Chemical trns-4-hydroxy L-ProlIn 25g
SDP
Supplier:  Cayman Chemical 2606125 G
Encompass_Preferred
A chiral building block; commonly used as a chiral building block in the organic synthesis of various compounds; a constituent of collagen and elastin and has been found in many bacterial secondary metabolites, including echinocandins
Catalog No. 50-293-6332