Learn More
Cayman Chemical trns-ClovamIde 5mg
SDP
Supplier:  Cayman Chemical 161385 MG
Encompass_Preferred
A naturally occurring caffeoyl conjugate with antioxidant and antiradical properties; demonstrates neuroprotective effects (EC50s = 0.9-3.7 µM) in several in vitro models of neuronal death
Catalog No. 50-297-2310