Learn More
Cayman Chemical trns-Resveratrol 250mg
SDP
Supplier:  Cayman Chemical 70675250 MG
Encompass_Preferred
A polyphenol with diverse biological activities; inhibits the cyclooxygenase and hydroperoxidase activities of COX-1 (EC50s = 15 and 3.7 μM, respectively); inhibits ERK1, JNK1, Src, PKCα, aromatase/CYP19, and DNA polymerases α and δ (IC50s = 37, 50, 20, in vitro and/or ex vivo; exhibits anticancer, anti-inflammatory, and antiviral activities in mice
Catalog No. 50-289-6245