Learn More
Cayman Chemical trns-Resveratrol-d4 500ug
SDP
Supplier:  Cayman Chemical 13130500 UG
Encompass_Preferred
An internal standard for the quantification of trans-resveratrol by GC- or LC-MS
Catalog No. 50-295-5914