Learn More
Cayman Chemical trns-VaccenIc AcId 50mg
SDP
Supplier:  Cayman Chemical 1530150 MG
Encompass_Preferred
An ω-7 fatty acid; dietary administration reduces total body fat, mesenteric fat, and adipocyte size, increases inguinal fat mass, and decreases intestinal and hepatic triglyceride secretion in a rat model of obesity with features of metabolic syndrome at 1% w/w; decreases hepatocellular ballooning and steatosis in the same model; dietary administration of a butter enriched with trans-vaccenic acid decreases serum cholesterol levels and the formation of aortic atherosclerotic lesions in Ldlr-/- mice
Catalog No. 50-297-3063