Learn More
Cayman Chemical trnsFarnsen 500mg
SDP
Supplier:  Cayman Chemical 31502500 MG
Encompass_Preferred
A volatile sesquiterpene hydrocarbon with pheromonal and kairomonal activities; produced by aphids in response to predator or parasite attack; induces dispersion of 50% of M. persicae aphids from a cluster at 0.2 ng; attracts larvae and adults of A. bipunctata at >2 μg
Catalog No. 50-299-0971