Learn More
Cole-Parmer Argos Technologies Transfer Pipette, 23.0 mL, Extra long; 100/Dispenser Box
Supplier:  Cole-Parmer 0622624
Encompass
Low-density polyethylene (LDPE) makes these disposable pipettes with built-in bulbs the perfect substitute for breakable glass pipettes, eliminating cross-contamination and cumbersome rubber bulb changes. Plastic reduces the loss of cells and proteins that often occurs with binding that takes place with glass alternatives. Precision manufacturing ensures uniform repeatable drops. Heat seal pipettes to provide liquid-tight transport and storage. These pipettes are suitable for use with a temperature range of -50 to 90°C and can be gas sterilized.
Catalog No. 50-196-0570