Learn More
Cayman Chemical TrnsfRrIn RECPTR ProteIn
SDP
Supplier:  Cayman Chemical 37081100 TEST
Encompass_Preferred
Immunogen: Recombinant human TfR1 • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) TfR1 • Applications: FC
Catalog No. 50-291-4888