Learn More
Cayman Chemical TrnsportntrIfluoroacetate 10mg
SDP
Supplier:  Cayman Chemical 3749710 MG
Encompass_Preferred
A cell-penetrating peptide; has been used in the surface functionalization of liposomes for drug delivery in vitro and in vivo and for the delivery of fluorescent cargo in vitro
Catalog No. 50-291-5808