Learn More
Medchemexpress LLC HY-119824 5mg Medchemexpress, Trofosfamide CAS:22089-22-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1198245MG
Encompass_Preferred
Medchemexpress, HY-119824 5mg Trofosfamide CAS:22089-22-1 Trofosfamide is an orally bioavailable oxazaphosphorine derivative with antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9550