Learn More
Medchemexpress LLC HY-50935 100mg , Troglitazone CS-045 CAS:97322-87-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY50935100MG
Encompass_Preferred
Medchemexpress, HY-50935 100mg Troglitazone CS-045 CAS:97322-87-7 Formula:C24H27NO5S EC50: 550 nM (Human PPARγ), 780 nM (Murine PPARγ) Purity:98% Troglitazone is a PPARγ agonist, with EC 50 s of 550 nM and 780 nM for human and murine PPARγ receptor, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2799