Learn More
Medchemexpress LLC Troglitazone-d4 | 2749370-85-0 | 98.19% | C24H23D4NO5S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY50935S1MG
Encompass_Preferred
Troglitazone-d4 is deuterium labeled Troglitazone. Troglitazone is a PPARγ agonist, with EC50s of 550 nM and 780 nM for human and murine PPARγ receptor, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-58367