Learn More
Abcam Troglitazone, PPAR-gamma receptor agonist, 10MG
Supplier:  Abcam AB14111210MG
Encompass_Preferred
MW 441.5 Da, Purity >97%. Potent, selective PPAR-γ receptor agonist (EC₅₀ = 780 nM). No activity at PPAR-α or PPAR-δ (up to 10 mM). Orally active. Antidiabetic and anticancer agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6895