Learn More
Apexbio Technology LLC Trometamol (Tris, Tromethamine) 77-86-1 50mg
SDP
Supplier:  Apexbio Technology LLC B186150
Encompass_Preferred
Trometamol (Tris Tromethamine) (CAS 77-86-1) is a small-molecule buffer system It is designed to stabilize and adjust pH in biological and biochemical environments by accepting protons and binding excess hydrogen ions Trometamol exerts its biological activity primarily through proton binding effectively buffering pH changes and normalizing extracellular pH Based on these properties Trometamol holds research potential in the stabilization of experimental solutions maintenance of pH during cell culture enzymatic reactions electrophoresis protein purification and studies on acid-base imbalance
Catalog No. 50-000-88565