Learn More
Medchemexpress LLC Trontinemab 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP999910MG
Encompass_Preferred
Trontinemab (RG6102) is a brain-penetrant anti-amyloid bispecific and humanizedized IgG1- antibody targeting to A plaques and transferrin receptor 1 (TFR1) Trontinemab binds to fibrillar A as well as A plaques triggering plaque clearance by engaging immune cells on Alzheimer disease (AD) brain sections Trontinemab also shows specific affinity to cynomolgus and human TFR1[1][2][3][4]
Catalog No. 50-003-21081