Learn More
Medchemexpress LLC TROP-2 Cynomolgus 500ug | 500ug
SDP
Supplier:  Medchemexpress LLC HYP77861500UG
Encompass_Preferred
The TROP-2 protein emerged as a potential growth factor receptor implying involvement in cellular processes related to growth and signaling As a putative receptor TROP-2 may play a crucial role in transducing signals that regulate cell growth and proliferation TROP-2 Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived TROP-2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87698