Learn More
Bioss Trop-2/TACSTD2 Extracellular
SDP
Supplier:  Bioss BSM70695M
Encompass_Preferred
Trop-2/TACSTD2 (Extracellular region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M005Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65720