Learn More
Medchemexpress LLC Tropicamide | 1508-75-4 | 98.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB03211G
Encompass_Preferred
Tropicamide is a selective M4 muscarinic acetylcholine receptor antagonist. It is commonly used as an eye drop to induce short-term mydriasis (pupil dilation) and cycloplegia. This compound is valuable in the research of neurological diseases, such as epilepsy.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Improves perseverative behavior, learning, and memory in Fmr1KO mice models
  • Reduces the occurrence of seizures
  • Decreases pilocarpine-induced tremulous jaw movements in rat models
Catalog No. 50-004-06895