Learn More
Medchemexpress LLC Tropicamide-d3 | 2673270-13-6 | 99.1% | 287.37 g/mol | C17H17D3N2O2 | 10 MG

Supplier: Medchemexpress LLC HYB0321S10MG
Tropicamide-d3 is a deuterium-labeled isotopologue of tropicamide supplied for analytical and research use as a high-purity reference standard. It is intended as an internal standard for mass spectrometry and other quantitative assays where isotopic labelling improves accuracy and tracking.
- High purity (99.1%).
- Deuterium-labeled internal standard for analytical mass spectrometry.
- Molecular weight 287.37 g/mol; molecular formula C17H17D3N2O2.
- CAS number 2673270-13-6.
- Soluble in DMSO, ethanol, and DMF.
- Supplied as a small vial (10 mg) suitable for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.