Learn More
Medchemexpress LLC Tropicamide-d3 | 2673270-13-6 | 99.1% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0321S5MG
Encompass_Preferred
Tropicamide-d3 is the deuterium labeled form of Tropicamide. Tropicamide is a selective M4 muscarinic acetylcholine receptor antagonist that produces short-acting mydriasis and cycloplegia when applied as eye drops. Deuterium substitution in drug molecules is of interest due to its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06894