Learn More
Medchemexpress LLC Tropifexor | 1383816-29-2 | MFCD31693060 | 99.4% | 603.6 g/mol | C29H25F4N3O5S | 200 MG
SDP
Supplier:  Medchemexpress LLC HY107418200MG
Encompass_Preferred
Tropifexor is a non-bile-acid, small-molecule agonist of the farnesoid X receptor (FXR) with reported biochemical potency around EC50 ≈ 0.2 nM. Identified in medicinal chemistry studies and advanced into clinical development, it is used in research on cholestatic liver diseases and nonalcoholic steatohepatitis (NASH). Available as a research reagent with reported high purity and defined analytical properties.
  • Highly potent FXR agonist (EC50 ≈ 0.2 nM).
  • Non-bile-acid small molecule suitable for biochemical and cellular assays.
  • Reported high purity for research applications.
  • Well characterized chemical properties for analytical workflows.
  • Supported by published discovery and clinical development literature.
Catalog No. 50-002-41406