Learn More
Selleck Chemical LLC Tropifexor S8733-2MG
SDP
Supplier:  Selleck Chemical LLC S87332MG
Encompass_Preferred
Tropifexor (LJN452) is a novel and highly potent agonist of FXR with EC50 of 0 2 nM in HTRF assay It shows no significant off-target activity against a broad panel of enzyme ion channel nuclear receptor and GPCR ( 10000-fold selectivity for FXR)
Catalog No. 50-000-24568