Learn More
Medchemexpress LLC Tropinone | 532-24-1 | MFCD00005549 | 100.0% | 139.20 g/mol | C8H13NO | 25g
SDP
Supplier:  Medchemexpress LLC HYY013525G
Encompass_Preferred
Tropinone is an organic synthesis intermediate and a skeleton for constructing derivatives containing thiazole rings and other structures Tropinone derivatives have anticancer activities such as inducing tumor cell apoptosis and inhibiting tyrosinase activity and can be used in research such as anticancer drug development and tyrosinase inhibitor design[1]
Catalog No. 50-002-69847