Learn More
Medchemexpress LLC Tropisetron Hydrochloride | 105826-92-4 | 100.0% | C17H21ClN2O2 | 5 MG
Supplier: Medchemexpress LLC HYB00205MG
Tropisetron Hydrochloride is a neuroprotective agent that functions as a 5-HT3 receptor antagonist, a calmodulin inhibitor, and an α7-nAChR receptor agonist, with an IC50 of 70.1 nM for the 5-HT3 receptor. It exhibits anti-inflammatory properties and immune-regulating functions, helping to alleviate symptoms associated with chemotherapy and post-surgery. Additionally, it has been shown to reduce Ab-induced hippocampal neuroinflammation.
- Acts as a 5-HT3 receptor antagonist
- Functions as a calmodulin inhibitor
- Acts as an α7-nAChR receptor agonist
- Exhibits anti-inflammatory properties
- Possesses immune-regulating functions
- Helps alleviate symptoms associated with chemotherapy and post-surgery
- Reduces Ab-induced hippocampal neuroinflammation