Learn More
Apexbio Technology LLC Tropisetron Hydrochloride 105826-92-4 5g
SDP
Supplier:  Apexbio Technology LLC B22585000
Encompass_Preferred
Tropisetron Hydrochloride is an antagonist targeting serotonin type 3 (5-HT3) receptors and an agonist at 7-nicotinic acetylcholine receptors ( 7nAChR) It is designed to modulate serotonergic and cholinergic signaling thereby regulating neuronal and gastrointestinal pathways Tropisetron Hydrochloride exerts its biological activity primarily through specific binding to and inhibition of 5-HT3 receptor activity It demonstrates inhibitory activity with an IC50 value of approximately 70 nM Based on these pharmacological properties Tropisetron Hydrochloride holds research potential in neuropharmacology neuroscience gastroenterology and oncology for investigating serotonin-mediated neuronal signaling gastrointestinal motility regulation chemotherapy-induced nausea and vomiting and cholinergic modulation via 7nAChR
Catalog No. 50-000-89881