Learn More
Medchemexpress LLC Tropolone | 533-75-5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN713510MM/1ML
Encompass_Preferred
Tropolone is a seven-membered non-benzenoid aromatic compound, serving as a precursor for various Azulene derivatives. It is recognized as a potent mushroom tyrosinase inhibitor, with an IC50 of 0.4 μM, and its inhibitory effects are reversible. This compound exhibits broad anti-viral and anti-fungal activities, demonstrating synergistic effects when co-treated with nucleos(t)ide analog drugs, and shows promise for osteosarcoma research.
  • Potent mushroom tyrosinase inhibitor
  • Reversible inhibitory effect
  • Broad anti-viral and anti-fungal activity
  • Synergistic with nucleos(t)ide analog drugs
  • Promising for osteosarcoma research
Catalog No. 50-004-18128