Learn More
Medchemexpress LLC Tropolone | 533-75-5 | 25 G
Supplier: Medchemexpress LLC 25GHYN7135
Tropolone is a seven-membered non-benzenoid aromatic compound and a precursor of many Azulene derivatives. It is a potent mushroom tyrosinase inhibitor with an IC50 value of 0.4 μM. Tropolone also shows broad anti-viral and anti-fungal activity and is synergistic when co-treated with nucleos(t)ide analog drugs. It is a promising candidate for research in osteosarcoma.
- Potent mushroom tyrosinase inhibitor (IC50: 0.4 μM)
- Exhibits broad anti-viral and anti-fungal activity
- Synergistic upon co-treatment with nucleos(t)ide analog drugs
- Promising candidate for research in osteosarcoma