Learn More
Medchemexpress LLC Tropolone | 533-75-5 | MFCD00004158 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN71351G
Encompass_Preferred
Tropolone is a seven-membered non-benzenoid aromatic compound that serves as a precursor for many Azulene derivatives. It is a potent mushroom tyrosinase inhibitor with an IC50 value of 0.4 μM. Tropolone exhibits broad anti-viral and anti-fungal activity and is considered a promising candidate for osteosarcoma research.
  • Potent mushroom tyrosinase inhibitor with an IC50 value of 0.4 μM
  • Exhibits broad anti-viral and anti-fungal activity
  • Shows synergistic effects when co-treated with nucleos(t)ide analog drugs
  • Considered a promising candidate for osteosarcoma research
  • Induces apoptotic cell death in a variety of human osteosarcoma cell lines
Catalog No. 50-004-19879