Learn More
Medchemexpress LLC Tropolone | 533-75-5 | MFCD00004158 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN71355G
Encompass_Preferred
Tropolone is a seven-membered non-benzenoid aromatic compound that acts as a precursor for many Azulene derivatives. It is a potent mushroom tyrosinase inhibitor (IC50 0.4 μM) and demonstrates broad anti-viral and anti-fungal activity. Tropolone is considered a promising candidate for research in osteosarcoma.
  • Seven-membered non-benzenoid aromatic compound
  • Precursor for Azulene derivatives
  • Potent mushroom tyrosinase inhibitor (IC50 0.4 μM)
  • Exhibits broad anti-viral and anti-fungal activity
  • Shows synergy with nucleos(t)ide analog drugs
  • Promising candidate for osteosarcoma research
  • Induces apoptotic cell death in human osteosarcoma cell lines
  • Disrupts osteosarcoma cell migration and invasion
Catalog No. 50-004-17818