Learn More
Medchemexpress LLC Tropolone | 533-75-5 | MFCD00004158 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN713550G
Encompass_Preferred
Tropolone is a seven-membered non-benzenoid aromatic compound known for its diverse biological activities and chemical applications. It functions as a potent mushroom tyrosinase inhibitor and exhibits broad anti-viral and anti-fungal properties. Tropolone also serves as a sensitive reagent for reducing sugars and an organic chelator, capable of forming complexes with trivalent lanthanide ions.
  • Potent mushroom tyrosinase inhibitor
  • Exhibits broad anti-viral and anti-fungal activity
  • Sensitive reagent for reducing sugars
  • Organic chelator for trivalent lanthanide ions
  • Precursor for synthesizing azulene derivatives
  • Used in the preparation of fused heterocycles and complexes of Ga(III) and In(III)
  • Promising candidate for osteosarcoma research
Catalog No. 50-004-21094