Learn More
Medchemexpress LLC Tropone (cycloheptatrienone) | 539-80-0 | MFCD00014331 | 97.0% | 106.12 g/mol | C7H6O | 1 ML

Supplier: Medchemexpress LLC HYW03590410MM/1ML
Tropone (cycloheptatrienone) is a non-benzenoid aromatic compound used as a chemical building block and drug intermediate. It is commonly used as a precursor in the synthesis of tropolone and other tropane-type natural products, and is supplied in solution and bulk forms with accompanying analytical documentation for research and development work.
- Non-benzenoid aromatic building block used in organic synthesis.
- Precursor to tropolone and related natural products.
- Available as a 10 mM solution in DMSO and in multiple bulk sizes.
- Provided with supporting documents including COA, SDS, HNMR, and GC.
- Molecular formula C7H6O and molecular weight 106.12 g/mol.
- CAS number 539-80-0 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.