Learn More
Medchemexpress LLC Tropone (cycloheptatrienone) | 539-80-0 | MFCD00014331 | 97.8% | 106.12 g·mol⁻1 | C7H6O | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW035904500MG
Encompass_Preferred
Tropone (cycloheptatrienone) is a seven-membered non-benzenoid aromatic ketone used as a chemical intermediate and reagent in organic synthesis. It is commonly employed in the preparation of tropolone derivatives and other tropane-type natural product intermediates, and is supplied with documented physical and purity specifications for laboratory research use.
  • Seven-membered non-benzenoid aromatic ketone useful as a chemical intermediate.
  • Documented purity: 97.8% (specific batches reported up to 99.48%).
  • Offered in a 500 mg package size suitable for small-scale research.
  • Defined physical properties: molecular weight 106.12 g·mol⁻1 and formula C7H6O.
  • Provided with supporting documentation including data sheet, COA, and SDS.
  • Storage and shipping conditions specified for both pure material and solvent solutions.
Catalog No. 50-002-99536