Learn More
Medchemexpress LLC Trospium chloride | 10405-02-4 | 99.93% | 1 ML

Supplier: Medchemexpress LLC HYB046110MM/1ML
Trospium chloride is an orally active, specific, and competitive antagonist of muscarinic cholinergic receptors (mAChRs), exhibiting antimuscarinic activity. It binds with high affinity to muscarinic receptors M1, M2, and M3, but not to nicotinic cholinergic receptors.
- CAS No.: 10405-02-4
- Molecular weight: 427.96
- Formula: C25H30ClNO3
- Appearance: Solid
- Color: White to off-white
- Purity: 99.93%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.