Learn More
Medchemexpress LLC Trovirdine | 149488-17-5 | 99.4% | 337.24 | C13H13BrN4S | 10 MG

Supplier: Medchemexpress LLC HY1534910MG
Trovirdine (LY300046) is a small-molecule inhibitor of HIV-1 reverse transcriptase provided as a research reagent. It is intended for in vitro biochemical and anti-infection studies and is supplied in multiple pack sizes and formulations; detailed purity, handling, and storage information is provided by the manufacturer.
- Inhibits HIV-1 reverse transcriptase in biochemical assays.
- Reported purity approximately 99.4%.
- Molecular formula C13H13BrN4S and molecular weight 337.24 g/mol.
- Available in small milligram quantities and as a DMSO solution for reconstitution.
- Recommended storage: powder and in-solvent conditions per manufacturer guidance.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.