Learn More
Apexbio Technology LLC Troxipide 30751-05-4 50mg
SDP
Supplier:  Apexbio Technology LLC B186250
Encompass_Preferred
Troxipide is a gastric cytoprotective compound It is designed to promote mucus secretion enhance endogenous prostaglandin production and facilitate mucosal barrier repair thereby supporting gastrointestinal mucosal protection and regeneration Troxipide exerts its biological activity primarily through mucosal protective anti-ulcer and anti-inflammatory mechanisms independent of gastric or duodenal pH and without suppressing gastric acid secretion Based on these pharmacological properties Troxipide holds research potential in investigating gastrointestinal mucosal protection mechanisms mucosal regeneration pathways and inflammatory modulation in gastrointestinal disorders including ulcers and inflammatory mucosal injury models
Catalog No. 50-000-92717