Learn More
CPC Scientific H-Trp-Asn-Arg-Gln-Leu-Tyr-Pro-Glu-Trp-Thr-Glu-Ala-Gln-Arg-Leu-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANS009B
Encompass
ONE-LETTER SEQUENCE: WNRQLYPEWTEAQRLDMOLECULAR FORMULA: C95H137N27O28MOLECULAR WEIGHT:2105.27STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [195523-86-5], SYNONYMS: Melanocyte Protein PMEL 17 (44-59) (human, bovine, mouse), Melanocyte Lineage-Specific Antigen GP100 (44-59) (human, bovine, mouse), Melanoma-Associated ME20 Antigen (44-59) (human, bovine, mouse), ME20M/ME20S (44-59) (human, bovine, mouse), ME20-M/ME20-S (44-59) (human, bovine, mouse), 95 kDa Melanocyte-Specific Secreted Glycoprotein (44-59) (human, bovine, mouse)RESEARCH AREA: Cancer
Catalog No. 50-210-8912