Learn More
ABclonal Technology TRPC1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1252520UL
Encompass_Preferred
The protein encoded by this gene is a membrane protein that can form a non-selective channel permeable to calcium and other cations. The encoded protein appears to be induced to form channels by a receptor tyrosine kinase-activated phosphatidylinositol second messenger system and also by depletion of intracellular calcium stores. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-14880