Learn More
CPC Scientific H-Thr-Arg-Pro-Ile-Ile-Thr-Thr-m-nitro-Tyr-Gly-Pro-Ser-Asp-Lys(Abz)-Tyr-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HRV14002B
Encompass
This fluorogenic (FRET) substrate containing an anthraniloyl group and a 3-nitrotyrosine as the resonance energy transfer donor/quencher pair, was developed for the assay of 2A protease from human rhinovirus (kcat/Km = 154 M-1s-1), which has been viewed as an important enzyme target for antiviral intervention.ONE-LETTER SEQUENCE: TRPIITT-m-nitro-YGPSD-K(Abz)-YMOLECULAR FORMULA: C80H118N20O26MOLECULAR WEIGHT:1775.94STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [213135-91-2], RESEARCH AREA: AntimicrobialREFERENCES: Q.M.Wang et al., Arch. Biochem. Biophys., 356, 12 (1998)
Catalog No. 50-210-9578