Learn More
Medchemexpress LLC TRPM7-IN-1 | 1909225-07-5 | 98.8% | 419.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1686085MG
Encompass_Preferred
TRPM7-IN-1 (compound SUD), a benzoylurea derivative, is an effective TRPM7 inhibitor. It induces cell cycle arrest and apoptosis, and decreases the migration of MCF-7 and BGC-823 cells. It also decreases vimentin expression and increases E-cadherin expression. This compound potentially reduces TRPM7-like current and decreases TRPM7 expression through the PI3K/Akt signaling pathway, making it a potential agent to suppress the metastasis of breast and gastric cancer by inhibiting TRPM7 expression and function.
  • Effective TRPM7 inhibitor.
  • Induces cell cycle arrest and apoptosis.
  • Decreases migration of MCF-7 and BGC-823 cells.
  • Decreases vimentin expression and increases E-cadherin expression.
  • Potentially reduces TRPM7-like current and decreases TRPM7 expression via the PI3K/Akt signaling pathway.
  • Potential agent to suppress metastasis of breast and gastric cancer.
Catalog No. 50-003-90421