Learn More
Meridian Life Science Tru Block, 0.1 g
SDP
Supplier:  Meridian Life Science A66800H0.1
Encompass
HAMA Blocker against Immunoassay Interference - actively blocks Heterophilic Antibody (HA) interference in immunoassays including, but not limited to, Human Anti-Mouse Antibody (HAMA) interference.
Catalog No. NC1003610