Learn More
Biotium TrueBlack IF Background Suppressor System (Permeabilizing)
Supplier:  Biotium 23012T
Encompass_Preferred
The TrueBlack Background Suppressor System (Permeabilizing) is a buffer system designed for blocking background from both non-specific antibody binding as well as direct interaction of fluorescent dyes on antibodies with cells or tissue sections.
Catalog No. 50-196-4406