Learn More
Rockland Immunochemicals TrueBlot Anti-Mouse Ig IP Agarose Beads - 00-8811-25
SDP
Supplier:  Rockland Immunochemicals 00881125
Encompass
TrueBlot Anti-Mouse Ig IP Agarose Beads are a suspension of activated agarose beads coupled with goat Anti-mouse IgG. It is suitable for precipitation of mouse IgGs used as the primary antibodies in immunoprecipitation assays. The beads are in suspension and will settle upon storage. Prior to use, mix the vial gently (do not vortex) to ensure delivery of proper bead volume.
Catalog No. NC0421973