Learn More
MSC Trumeter Length Counter, 18 in Knurled Aluminum Wheel, Full Feet, Top Coming
Supplier:  MSC 07424898
Encompass
This heavy-duty mechanical length measuring unit is designed for continuous, high-speed operation and long-lasting performance. Featuring an 18 in knurled aluminum wheel, it measures in full feet with a top coming direction of travel. Its twin wheel design ensures stability, and a stay rod mounting allows for pivoting and easy lifting away from the material.
  • Large 6-digit display with reset
  • Constructed from die-cast aluminum with PC-ABS plastic base and ABS plastic cover, offering IP40 protection
  • Measures up to 3,000 rpm with +/- 1% accuracy
  • Can be mounted or hung using the supplied stay rod
  • Weighted design maintains contact with the material being measured
  • 7 mm diameter shaft with double oil-impregnated bronze bearings
  • Lever reset option
  • Suitable for a wide range of continuous manufacturing processes including textile, paper, plastic film, and metal processing
Catalog No. 50-005-29560