Learn More
AdipoGen Trypacidin
SDP
Supplier:  AdipoGen BVT0442M001
Encompass_Preferred
Chemical. CAS 1900-29-4. Formula C18H16O7. MW 344.3. Isolated from Aspergillus fumigatus. Antibiotic. Antibacterial and antiprotozoal trypanocidal. Toxic to human A549 lung cells. Necrosis inducer. Toxic to human bronchial epithelial cells. Stimulating the growth of corn seedling roots. Inhibits IL-6 secretion, collagen IV and fibronectin production
Catalog No. 50-256-1924