Learn More
Alkali Scientific Trypan Blue, 500 G
SDP
Supplier:  Alkali Scientific RYP004CP
Encompass_Preferred
Trypan Blue, 500g is an essential dye for staining dead cells in cell viability assays. When applied, it selectively stains cells with compromised membranes, indicating cell death. This dye is useful in numerous laboratory applications, especially in cytotoxicity testing, where it helps determine the effects of different substances on cell health. The 500g pack size is ideal for laboratories with high throughput needs, such as those running large-scale experiments or requiring multiple uses for continuous testing. Trypan Blue is widely used in clinical research, drug development, and toxicology studies to measure cell viability and assess cellular damage in various treatment scenarios.
Catalog No. 50-006-61784