Learn More
Worthington Biochemical Corporation Trypsin, 1g
SDP
Supplier:  Worthington Biochemical Corporation LS003703
Encompass
Supplied as a dialyzed and lyophilized powder. Store at 2-8°C. Protect from moisture.
Catalog No. NC9811754