Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin Sequencing Grade, 4 x 25 ug
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11418475001
Encompass
Trypsin Sequencing Grade is used to to digest proteins in solution, in gels or on blotting membranes.The enzyme is used for protein-structure elucidation, tryptic mapping, fingerprinting, sequence analysis, and translocation studies. Trypsin Sequencing Grade generates glycopeptides from purified glycoproteins and is suited for the digestion of proteins in polyacrylamide gels.
Catalog No. NC1941352