Learn More
Cayman Chemical TryptamIn-d4hydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 335381 MG
Encompass_Preferred
An internal standard for the quantification of tryptamine by GC- or LC-MS
Catalog No. 50-295-0946