Learn More
Selleck Chemical LLC Tryptamine S3627-1g
SDP
Supplier:  Selleck Chemical LLC S36271G
Encompass_Preferred
Tryptamine is a monoamine alkaloid and may play a role as a neuromodulator or neurotransmitter It is an agonist of hTAAR1 It acts as a non-selective serotonin receptor agonist and serotonin-norepinephrine-dopamine releasing agent (SNDRA)
Catalog No. 50-311-0755