Learn More
Medchemexpress LLC Tryptamine | 61-54-1 | MFCD00005661 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB21325G
Encompass_Preferred
Tryptamine is a selective, blood-brain-penetrating 5-HT4 receptor agonist and an endogenous ligand of the aryl hydrocarbon receptor (AHR). It promotes intestinal anion secretion and fluid transport by activating G protein-coupled receptors (GPCRs) and accelerates gastrointestinal motility. Tryptamine can be used to study intestinal motility disorders such as functional constipation and has shown significant efficacy in multiple sclerosis models.
  • Regulates Th17/Treg balance to inhibit neuroinflammation
  • Competitively binds to 5-HT receptors to regulate central nervous system activity
  • Participates in temperature regulation and spinal reflex regulation as a neuromodulator
Catalog No. 50-004-09909